Anti-CMV-p65 Mouse Monoclonal Antibody (CF640R) [clone: CMV100]
Leverandør: Biotium
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BNC400100-500 BNC400100-100
BNC400100-500EA 4180 DKK
BNC400100-500 BNC400100-100
Anti-CMV-p65 Mouse Monoclonal Antibody (CF640R) [clone: CMV100]
Antistoffer
Ten to forty percent of the patients with acquired immunodeficiency syndrome (AIDS) develop cytomegalovirus (CMV) infections. In some patients with AIDS, CMV is detected in the bronchoalveolar lavage fluid (BALF), urine, and other specimens, even when there are no symptoms of CMV disease. An indicator of active CMV infection is needed to facilitate the diagnosis of CMV disease in patients with AIDS or HIV infection. CMV p65 antigen was detected in the leukocytes of both the peripheral blood and BALF during the early phase of CMV disease.
Type: Primary
Antigen: Not Applicable
Clonality: Monoclonal
Clone: CMV100
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: CMV