Anti-CMV-p65 Mouse Monoclonal Antibody [clone: CMV100]

Leverandør: Biotium
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BNUB0100-500 BNUB0100-100
BNUB0100-500EA 3810 DKK
BNUB0100-500 BNUB0100-100
Anti-CMV-p65 Mouse Monoclonal Antibody [clone: CMV100]
Antistoffer
Ten to forty percent of the patients with acquired immunodeficiency syndrome (AIDS) develop cytomegalovirus (CMV) infections. In some patients with AIDS, CMV is detected in the bronchoalveolar lavage fluid (BALF), urine, and other specimens, even when there are no symptoms of CMV disease. An indicator of active CMV infection is needed to facilitate the diagnosis of CMV disease in patients with AIDS or HIV infection. CMV p65 antigen was detected in the leukocytes of both the peripheral blood and BALF during the early phase of CMV disease.

  • Purified Antibody

Type: Primary
Antigen: CMV-p65
Clonality: Monoclonal
Clone: CMV100
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR