Anti-MAGEA1 Mouse Monoclonal Antibody (CF568) [clone: MA454]

Leverandør: Biotium
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BNC680008-500 BNC680008-100
BNC680008-500EA 4180 DKK
BNC680008-500 BNC680008-100
Anti-MAGEA1 Mouse Monoclonal Antibody (CF568) [clone: MA454]
Antistoffer
Recognizes a protein of 42-46 kDa, identified as MAGE-1. This MAb does not cross-react with MAGE-2, -3, -4, -6 -9, -10, -or -12 protein. Human malignant neoplasms carry rejection antigens that are recognized by the patients' autologous, tumor directed and specific, cytolytic, CD8 T lymphocyte clones (CTL). The MAGE family of genes codes an important group of antigens. It was identified that melanomas and primary glial brain tumors express common melanoma associated antigens (MAAs). Because MAGE-1 is expressed on a significant proportion of human neoplasms of various histological types (melanoma, brain tumors of glial origin, neuroblastoma, non-small cell lung cancer, breast, gastric, colorectal, ovarian, renal cell carcinomas) and not on normal tissues, the encoded antigen may serve as a marker of early detection and target for cancer immunotherapy.

Type: Primary
Antigen: MAGEA1
Clonality: Monoclonal
Clone: MA454
Conjugation: CF568
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Dog, Human, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR