VWR® ARISTAR®, Titanium Standard Solutions for ICP-MS
Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
456962WEA 1240 DKK
456962W 85636.180
VWR® ARISTAR®, Titanium Standard Solutions for ICP-MS
Standarder Kationiske standarder Titanium Standards
  • Produced from high purity acids, water ASTM I 18 Megaohms
  • Solution assayed by titration
  • Final concentration verified against reference material from NIST
  • Total maximum uncertainty: 0.5% (typical)
  • Delivered with complete certificate of analysis
Certificeringer: Manufactured and tested in ISO 17034 / ISO 17025 accredited facilities