VWR® ARISTAR®, Beryllium Standard Solutions for ICP-MS
Leverandør: VWR Chemicals
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
456662NEA
2020
DKK
456662N
85553.180
VWR® ARISTAR®, Beryllium Standard Solutions for ICP-MS
Standarder
Kationiske standarder
Beryllium Standards
- Produced from high purity acids, water ASTM I 18 Megaohms
- Solution assayed by titration
- Final concentration verified against reference material from NIST
- Total maximum uncertainty: 0,5% (typical)
- Delivered with complete certificate of analysis
Certificeringer: Traceable to SRM from NIST. Manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.