Capillary columns, MXT®-200 (Siltek®-treated stainless steel)
Leverandør: Restek
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
75038 75052 75053 75055 75058 75070 75073 75082 75085 75088
REST75038EA 6740 DKK
REST75038 REST75052 REST75053 REST75055 REST75058 REST75070 REST75073 REST75082 REST75085 REST75088
Capillary columns, MXT®-200 (Siltek®-treated stainless steel)
Kromatografikolonner
MXT®-200 columns are general purpose columns for solvents, Freon® fluorocarbons, alcohols, ketones, silanes, and glycols.
  • Midpolarity phase; Crossbond® trifluoropropylmethyl polysiloxane
  • Temperature range up to 400 °C
  • Equivalent to USP G6 phase
Coil diameter: 114,3 mm