TwiXS packs, Acura® manual XS 826
Leverandør: Socorex Isba
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
826.TWX.N 826.TWX.E 826.TWX.G 826.TWX.K 826.TWX.H 826.TWX.Q
613-6875EA 3980 DKK
613-6875 613-6871 613-6872 613-6874 613-6873 613-6876
TwiXS packs, Acura® manual XS 826
Pipetter Automatiske pipetter
These ready-to-use kits include two Acura® manual XS pipettes, one shelf pipette holder, Qualitips® samples, QC certificates and operating instructions.