VWR® VWB2, Water Baths
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
462-0554EA 6290 DKK
462-0554 462-0555 462-0556 462-0557 462-0558 462-0559
VWR® VWB2, Water Baths
Bade Vandbade, uden omrøring
Unstirred water baths with double walled, stainless steel (inner) and coated steel sheet (outer) housing. All baths except the 2 litre models feature a drain tap for convenient emptying and are suitable for use with water or alternatively with heat transfer beads. Baths with a capacity of 5 litres or more have a heat transfer bead mode which is activated through the main menu. A drain stopper (plug) is supplied as on optional accessory to ensure heat transfer beads do not get stuck in the drain outlet. Note: Baths are classified Safety Class 1, allowing unattended operation when used with non flammable liquids.
  • Fast heat up, accurate temperature control
  • Digital temperature indicator with three temperature presets
  • Temperature range ambient 5 to 99 °C
  • Temperature stability ±0,2 °C
  • Adjustable over-temperature alarm protects samples from over heating
  • Dry run protection turns the bath off if no water or low water level is detected
  • Countdown timer and audible alarm: 1 to 999 minutes
Leveringsinformation: Supplied with a hinged clear polycarbonate lid (the 2 litre models have an unhinged lid) and universal tray. A wide range of optional accessories are available.