Anti-Agrin Mouse Monoclonal Antibody [clone: Agr-33]

Leverandør: ENZO LIFE SCIENCES
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ADIAGR510D ADIAGR510F ADIAGR510J
ENZOADIAGR510DEA 2140 DKK
ENZOADIAGR510D ENZOADIAGR510F ENZOADIAGR510J
Anti-Agrin Mouse Monoclonal Antibody [clone: Agr-33]
Antistoffer
Agrin is an essential extracellular matrix component which promotes clustering of nicotinic acetylcholine receptors (nAChRs) and other proteins at the neuromuscular junction during development.

  • Dilution range: 10 µg/ml for Immunohistochemistry

Agrin, MuSK, and Rapsyn are all essential components for AChR aggregation, through an unknown mechanism. The C-terminal region of agrin is released into the medium, interacts with receptors on the muscle surface and induces AChR aggregation.

Type: Primary
Antigen: Agrin
Clonality: Monoclonal
Clone: Agr-33
Conjugation:
Epitope: C-Terminal
Host: Mouse
Isotype: IgG1
Reactivity: Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR