Total alkalinity test kit, ORION®
Leverandør: Thermo Orion
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
700010 700012 700011-WA
ORIO700010EA 3800 DKK
ORIO700010 ORIO700012 ORIO700011-WA
Total alkalinity test kit, ORION®
Miljø test kit
Two-step direct measurement of total alkalinity and pH using a pH electrode.
  • Saves time by eliminating additional equipment set-up, tedious titrations and calculations
  • Direct reading of total alkalinity from 0 to 200 ppm as CaCO₃, from the enclosed pH conversion chart
  • No interference from color or turbidity
Leveringsinformation: Kit comprises alkalinity reagent (475 ml) ,alkalinity standard/control (475 ml), total alkalinity conversion chart and application procedure No. 517, total alkalinity in natural waters.