Bottles, wide neck, clear, with GLS 80® thread and baffles, DURAN®
Leverandør: DWK Life Sciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
212863658 212864457 212865453
215-3875EA 278 DKK
215-3875 215-3876 215-3877
Bottles, wide neck, clear, with GLS 80® thread and baffles, DURAN®
Flasker Glasflasker
Wide neck bottles made of clear DURAN® borosilicate glass 3.3, with GLS 80® thread, have three vertical baffles into the inner surface, the swirling motion is disrupted, giving a much more effective mixing motion.
  • Bottles are fully autoclavable
  • Easy to read graduated volume scale
  • Very good temperature resistance
  • Complete with PP pouring ring and GLS 80® screw cap
The baffles improve 'top to bottom' circulation and produce greater radial mixing. The baffles also offer other advantages such as suppressing the formation of a central vortex and reducing 'dead' zones where mixing does not occur. Compatible with DURAN® GLS 80® magnetic stirrers, other magnetic stir bars, and with the full range of wide mouth GLS 80® connection cap systems for liquid addition or removal when mixing with stirring bars. The baffles are moulded into the glass side wall of the bottles and form a continuous, smooth inner surface making clean up easy.