Anti-HSP20 Rabbit Polyclonal Antibody
Leverandør: ENZO LIFE SCIENCES
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ADISPA796D ADISPA796F
ENZOADISPA796DEA 2170 DKK
ENZOADISPA796D ENZOADISPA796F
Anti-HSP20 Rabbit Polyclonal Antibody
Antistoffer
Hsp20 is a member of the family of small heat shock proteins (sHsps), with homology to Hsp25, Hsp27, and alphaB-crystallin.
  • Dilution range: Western blot (1:1,000, colorimetric)
  • Detects a band of ~20kDa by Western blot
Type: Primary
Antigen: HSP20
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Dog, Fish, Human, Mouse, Rat