ATC probes, stainless steel, Orion™
Leverandør: Thermo Orion
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
927007MD 928007MD
662-1537EA 4760 DKK
662-1537 662-1582
ATC probes, stainless steel, Orion™
Elektroder pH-elektroder
Automatic Temperature Compensation (ATC) probes for fast, accurate measurements in the lab or field. Automatically correct pH measurements for variation in electrode slope due to temperature change.
Relative accuracy: ±1 °C