Size exclusion chromatography columns, PL aquagel-OH™ analytical
Leverandør: VARIAN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
PL1120-6520 PL1149-1530 PL1149-6240 PL1149-6250 PL1149-6260 PL1149-6801 PL1149-6820 PL1549-5800 PL1549-5801
VARIPL1120-6520EA 30580 DKK
VARIPL1120-6520 VARIPL1149-1530 VARIPL1149-6240 VARIPL1149-6250 VARIPL1149-6260 VARIPL1149-6801 VARIPL1149-6820 VARIPL1549-5800 VARIPL1549-5801
Size exclusion chromatography columns, PL aquagel-OH™ analytical
Kromatografikolonner
These PL aquagel-OH™ analytical coluns have a highly stable matrix ensuring reliable separations, even with modified eluents.
  • Highly versatile for neutral, polar, anionic and cationic samples
  • Mixed columns cover a wide range of molecular weights, simplifying column selection
These series have a pH range of 2-10, compatibility with organic solvent up to 50% methanol, mechanical stability up to 140 bar and low column operating pressures.