Propidiumiodid, dark red powder
Leverandør: MP Biomedicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Farlig
Synonymer: 3,8-Diamino-5-(3-(diethylmethylammonio)propyl)-6-phenylphenanthridiniumdiiodid
0219545810 0219545825 0219545880 0219545883
ICNA0219545810EA 375 DKK
ICNA0219545810 ICNA0219545825 ICNA0219545880 ICNA0219545883
Propidiumiodid, dark red powder
Propidium iodide
Propidium iodide is a fuorescent stain for nucleic acids.
Cell membrane integrity excludes propidium iodide from staining viable and apoptotic cells. Propidium iodide may be used in flow cytometry to evaluate cell viability when used with other dyes that stain viable cells or cells that are early in the apoptosis process.
Propidium iodide (PI) binds to DNA by intercalating between the bases with little or no sequence preference and with a stoichiometry of one dye per 4 to 5 base pairs of DNA. PI also binds to RNA, necessitating treatment with nucleases to distinguish between RNA and DNA staining. Once the dye is bound to nucleic acids, its fluorescence is enhanced 20- to 30-fold, the fluorescence excitation maximum is shifted ~30 to 40 nm to the red and the fluorescence emission maximum is shifted ~15 nm to the blue. Although its molar absorptivity (extinction coefficient) is relatively low, PI exhibits a sufficiently large Stokes shift to allow simultaneous detection of nuclear DNA and fluorescein-labeled antibodies, provided the proper optical filters are used. Propidium iodide is suitable for fluorescence microscopy, confocal laser scanning microscopy, flow cytometry and fluorometry.
Soluble at 1 mg/ml in water.
Formel: C₂₇H₃₄I₂N₄
Molekylvægt: 668.4 g/mol
MDL Number: MFCD00011921
CAS-nummer: 25535-16-4
Bestil nu
Specifikationer Testresultater
Identity Test Passes
FTIR Conforms to Standard
UV/VIS: l max (water) 493 ± 4 nm