Ammonium 1-pyrrolidinecarbodithioate ≥95%, white crystalline powder

Leverandør: MP Biomedicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Pyrrolidinedithiocarbamate ammonium salt, 1-Pyrrolidinecarbodithioic acid, ammonium salt, Ammonium pyrrolidine-1-carbodithioate, Pyrrolidine-1-Dithiocarboxylic Acid Ammonium Salt, APCD

0215647025 0215647080
ICNA0215647025EA 723 DKK
ICNA0215647025 ICNA0215647080
Ammonium 1-pyrrolidinecarbodithioate ≥95%, white crystalline powder
Ammonium 1-pyrrolidinecarbodithioate

Pyrrolidinedithiocarbamate, ammonium salt is a complexing agent for the extraction of metal ions.


Pyrrolidinedithiocarbamate, ammonium salt is used for the extraction and pre-concentration of heavy metals with an organic solvent from aqueous solutions for atomic absorption spectroscopy.


Prevents induction of nitric oxide synthetase by inhibiting translation of NOS mRNA; induces apoptosis in rat smooth muscle cells and inhibits apoptosis in leukemia HL-60 cells.


Soluble in water (100 mg/ml with heating and vortexing - creates a tan suspension).

Formel: C₅H₁₂N₂S₂
Molekylvægt: 146.3 · 18.0 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00012720
CAS-nummer: 5108-96-3

Bestil nu

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR