Roseoflavin ≥97%, brown powder

Leverandør: MP Biomedicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

0215425210 0215425225 0215425280
ICNA0215425210EA 2430 DKK
ICNA0215425210 ICNA0215425225 ICNA0215425280
Roseoflavin ≥97%, brown powder
Roseoflavin

Roseoflavin is a weak antibacterial agent active against some gram-positive bacteria, especially Staphylococcus aureus, Sarcina latea, Bacillus cereus and Bacillus subtilis. Also exhibits antiriboflavin activity and inhibits the biosynthesis of riboflavin by a repressor mechanism.


Advarsel: Research Use Only (RUO). Protect from light.

Formel: C₁₈H₂₃N₅O₆
Molekylvægt: 405.411 g/mol
Opbevaringstemperatur: Køleskab
CAS-nummer: 51093-55-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR