Pyruvate kinase, MP Biomedicals
Leverandør: MP Biomedicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
0215199905 0215199925 0215199950 0215199980
ICNA0215199905EA 1730 DKK
ICNA0215199905 ICNA0215199925 ICNA0215199950 ICNA0215199980
Pyruvate kinase, MP Biomedicals
Enzymer
Dissolves readily at 5 mg/ml in 0,06 M Tris/HCI pH 7,4 to give a clear colorless solution.
  • Protein determined by biuret
That amount of enzyme causing the oxidation of one micromole of NADH per minute at 25 °C and pH 7,4.