Oliesyre ≥99%, clear, colourless liquid
Leverandør: MP Biomedicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer: (9Z)-9-Octadecenoic acid
0215178101 0215178105 0215178125
ICNA0215178101EA 533 DKK
ICNA0215178101 ICNA0215178105 ICNA0215178125
Oliesyre ≥99%, clear, colourless liquid
Oliesyre
Oleic acid is a mono-unsaturated omega-9 fatty acid created by dehydrogenation of stearic acid. It is then further dehydrogenated and elongated to create members of the n-9 family of fatty acids.
Oleic acid is used as emollient. Small amounts of oleic acid are used as an excipient in pharmaceuticals. It is used as an emulsifying or solubilizing agent in aerosol products. Oleic acid have been shown to be important for the growth and productivity of Chinese Hamster Ovary (CHO) cells. Functions of Oleic Acid in Cell Culture Systems: Long-term energy storage: energy derived from NADPH and ATP is stored in fatty acids. Fatty acids are esterified to a glycerol backbone to form a group of compounds known as mono-, di- and tri- glycerides (neutral fats). Energy is released when fatty acids are degraded.
Soluble in methanol (40 mg/ml - clear solution), chloroform, benzene or fixed and volatile oils. Insoluble in water.
Advarsel: Research Use Only (RUO).
Formel: C₁₈H₃₄O₂
Molekylvægt: 282.47 g/mol
Kogepunkt: 286
Smeltepunkt: 13…14 °C (lit.)
Flammepunkt: 180
Opbevaringstemperatur: Fryser
MDL Number: MFCD00064242
CAS-nummer: 112-80-1
Merck Index: 13,06898
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e5aecc5dce4d0318386a45d7adb7c5822631010a&vid=892a2df4ecab3679b06983b84cd27ad334ef961f&prid=web&clid=SAPDAM
Bestil nu
Specifikationer Testresultater
Identity Test Passes
Purity ≥99%
FTIR Conforms to Standard