Phenazin, yellow crystalline powder
Leverandør: MP Biomedicals
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
0210260501
0210260510
ICNA0210260501EA
333
DKK
ICNA0210260501
ICNA0210260510
Phenazin, yellow crystalline powder
Phenazine
Fluorescence: λex 426 nm; λem 471 nm in ethyl acetate (Lit.).
Fluorescence: λex 426 nm; λem 471 nm in ethyl acetate (Lit.).
Advarsel: Research Use Only (RUO)
Formel:
C₁₂H₈N₂ Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00005023 CAS-nummer: 92-82-0 |
Specifikationer Testresultater
Identity Test | Passes |
FTIR | Conforms to Standard |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...