Adeninsulfat Dihydat ≥99%, powder

Leverandør: MP Biomedicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 6-Aminopurine sulphate dihydrate, Adenine hemisulphate dihydrate, 7H-Purin-6-amine hemisulphate dihydrate, 9H-Purin-6-amine hemisulphate dihydrate

0210019501 0210019525
ICNA0210019501EA 395 DKK
ICNA0210019501 ICNA0210019525
Adeninsulfat Dihydat ≥99%, powder
Adeninsulfat Dihydat

Adenine is a purine nucleobase with a wide range of chemical and biochemical roles in vivo and in vitro. It is a regulatory molecule and a component of DNA, RNA, cofactors (NAD, FAD) and signaling molecules (cAMP).


Solubility (0.5% aq soln): Clear, Colorless Solution.

Formel: (C₅H₅N₅)₂·H₂SO₄·2H₂O
Molekylvægt: 404.37 g/mol
Smeltepunkt: 260 °C (lit.)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00213655
CAS-nummer: 6509-19-9

Bestil nu

Specifikationer Testresultater

Identity Test Passes
Purity 101.7% (UV)
FTIR Conforms to Standard
Moisture 8,60%
pH (0.5% aq soln) 3,04
UV/VIS: l max (water) 260.6 nm
OD250/OD260 0.78
OD280/OD260 0.113
Solubility (0.5% aq soln) Clear, Colorless Solution


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR