HPLC columns, Polaris™ Si-A
Leverandør: VARIAN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
A2003020X020 A2003030X020 A2003050X030 A2003050X046 A2003050X212 A2003100X020 A2003100X030 A2003100X046 A2003125X040 A2003125X046 A2003150X020 A2003150X021 A2003150X030 A2003150X040 A2003150X046 A2003250X020 A2003250X030 A2003250X040 A2003250X046 A2003250X212 A2004250X212 A2005020X020 A2005030X020 A2005050X020 A2005050X030 A2005050X046 A2005100X020 A2005100X030 A2005100X046 A2005150X020 A2005150X030 A2005150X046 A2005250X020 A2005250X030 A2005250X046
VARIA2003020X020EA 7940 DKK
VARIA2003020X020 VARIA2003030X020 VARIA2003050X030 VARIA2003050X046 VARIA2003050X212 VARIA2003100X020 VARIA2003100X030 VARIA2003100X046 VARIA2003125X040 VARIA2003125X046 VARIA2003150X020 VARIA2003150X021 VARIA2003150X030 VARIA2003150X040 VARIA2003150X046 VARIA2003250X020 VARIA2003250X030 VARIA2003250X040 VARIA2003250X046 VARIA2003250X212 VARIA2004250X212 VARIA2005020X020 VARIA2005030X020 VARIA2005050X020 VARIA2005050X030 VARIA2005050X046 VARIA2005100X020 VARIA2005100X030 VARIA2005100X046 VARIA2005150X020 VARIA2005150X030 VARIA2005150X046 VARIA2005250X020 VARIA2005250X030 VARIA2005250X046
HPLC columns, Polaris™ Si-A
Kromatografikolonner
Polaris™ silica is ideal for the rapid separation of low molecular weight compounds that are soluble in organic solvents.
  • Porous silica particles, 5 to 10 µm in diameter
  • Spherical particles with pore size of 180 Å
Separations on silica columns depend upon the difference in orientation, type and number of functional groups associated with the compounds in the sample.