Cryoboxes, B55/B75/B95
Leverandør: GLW
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
B55 B55B B55G B55R B55S B55Y BS55 BS75 B95 B95B B95G B95R B95S B95Y BS95 B75 B75Y B75R B75S B65
GLWAB55EA 117 DKK
GLWAB55 GLWAB55B GLWAB55G GLWAB55R GLWAB55S GLWAB55Y GLWABS55 GLWABS75 GLWAB95 GLWAB95B GLWAB95G GLWAB95R GLWAB95S GLWAB95Y GLWABS95 GLWAB75 GLWAB75Y GLWAB75R GLWAB75S GLWAB65
Cryoboxes, B55/B75/B95
Bokse Æsker til Kryorør
Cryoboxes made of PP, natural or coloured.
  • Autoclavable up to +121 °C and use in freezer down to –90 °C
  • Included clear lid offers a view of the contents without opening the cryo-box
  • Drainage holes in base for easy opening the box at low temperatures
  • For light sensitive samples please choose the colour Black/black
Suitable for 25 vials, arranged in 5×5 compartments with 21,6 mm diameter. Suitable to store up to 25 HPLC vials, GC vials, tubes or wide-neck bottles and other flat-bottomed vials fit in the grid. The numeric labeling helps to identify your samples.
B55-Series:
Generic tube size up to 53 mm height and 21,6 mm diameter.
B75-Series:
Generic tube size up to 78 mm height and 21,6 mm diameter.
B95-Series:
Generic tube size up to 93 mm height and 21,6 mm diameter.