HPLC columns, Polaris™ C8-A
Leverandør: VARIAN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
A2010050X020 A2010050X046 A2010100X020 A2010100X030 A2010100X046 A2010125X040 A2010150X020 A2010150X030 A2010150X040 A2010150X046 A2010250X030 A2010250X040 A2010250X046 A2010250X100 A2010250X212 A2011020X020 A2011030X030 A2011050X020 A2011050X021 A2011050X046 A2011075X046 A2011100X046 A2011150X020 A2011150X046 A2011250X020
VARIA2010050X020EA 8650 DKK
VARIA2010050X020 VARIA2010050X046 VARIA2010100X020 VARIA2010100X030 VARIA2010100X046 VARIA2010125X040 VARIA2010150X020 VARIA2010150X030 VARIA2010150X040 VARIA2010150X046 VARIA2010250X030 VARIA2010250X040 VARIA2010250X046 VARIA2010250X100 VARIA2010250X212 VARIA2011020X020 VARIA2011030X030 VARIA2011050X020 VARIA2011050X021 VARIA2011050X046 VARIA2011075X046 VARIA2011100X046 VARIA2011150X020 VARIA2011150X046 VARIA2011250X020
HPLC columns, Polaris™ C8-A
Kromatografikolonner
These Polaris™ phases have rapid high resolution separations based on ultra pure silica to which alkyl chains and polar modifications are attached.
  • Enhanced retention and peak shape of polar analytes
  • Low hydrophobic than Polaris C18-A
  • Ideal for polar samples
  • Faster overall analysis times
The combination of high phase density bonding, ultra pure silica, and silanol shielding leads to excellent peak shape among polar-modified columns.