Hydrometers
Leverandør: GERING
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
0082 0083 0084 0085 0086 0087 0088 0089 0090 0091 0092 0093 0094 0095 0096
GERI0082EA 489 DKK
GERI0082 GERI0083 GERI0084 GERI0085 GERI0086 GERI0087 GERI0088 GERI0089 GERI0090 GERI0091 GERI0092 GERI0093 GERI0094 GERI0095 GERI0096
Hydrometers
Flydevægte
Set of 14 hydrometers calibrated at 20 °C.
Leveringsinformation: Supplied in a velvet lined case.