Cover glasses, square
Leverandør: GLASWARENFABRIK KARL HECHT
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
41000109 40990103 40990121 40990109 41000115 41000121 40990115 40990101
HECH41000109EA 314 DKK
HECH41000109 HECH40990103 HECH40990121 HECH40990109 HECH41000115 HECH41000121 HECH40990115 HECH40990101
Cover glasses, square
Dækglas
Borosilicate glass coverslips for histology and microscopy.