Air probes, clamp-type, for Curvex datalogger
Leverandør: Industrial Physics
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
CX2026 CX2022 CX2021 CX2020
SIMECX2026EA 2300 DKK
SIMECX2026 SIMECX2022 SIMECX2021 SIMECX2020
Air probes, clamp-type, for Curvex datalogger
Temperatur prober
These clamp-type probes can be used to measure air temperature.
  • Max. object thickness: 20 mm
  • Energy and/or water efficient
  • Recycled/renewable content