Adapters, tubing, KA, KECK™, DURAN®
Leverandør: DWK Life Sciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
286344006 286344109 286344203 286344306
271-7141EA 85.8 DKK
271-7141 271-7142 271-7145 271-7146
Adapters, tubing, KA, KECK™, DURAN®
Fittings og stik
KECK™ adapters are made up of glass-fibre reinforced, thermoplastic polyester (PBT) for glass hose couplings KA.
  • Protect fingers and hands from being injured
  • Prevents the breakage of glass hose couplings
  • Connect or remove hose by twisting ¼ turn
  • The clip, O-ring and the screwon cap are screwed onto the hose coupling permanently
Hose can be connected quickly, safely and without leakage (upto 30 bar pressure, 0,66 mbar vacuum).
Leveringsinformation: The adapters are available as a complete set which comes either with 4,5 / 9,0 mm Ø tubing part.