Multi-channel pipettes, mechanical, variable volume, F1-ClipTip™
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
4661120N 4661130N 4661140N 4661160N 4661170N 4661180N 4661210N 4661220N
613-5531EA 0 DKK
613-5531 613-5532 613-5533 613-5535 613-5536 613-5537 613-7150 613-7151
Multi-channel pipettes, mechanical, variable volume, F1-ClipTip™
Pipetter Automatiske pipetter
The Thermo Scientific™ F1-ClipTip™ pipette features interlocking technology to ensure secure tip attachment.
  • Silent and lighter volume adjustment mechanism with over 50% lower forces compared to earlier models
  • Improved grip on the volume adjustment knob ensures effortless and precise adjustment
  • Exclusive interlock design ensures each tip is clipped securely on the F1-ClipTip until it is released with a light touch
  • Secure volume setting prevents undesirable volume drift during operation
  • 120° adjustable finger rest – for both right- and left-handed operator comfort – allows the hand to relax during pipetting cycles
  • Lightweight handle design combined with low plunger, tip attachment and ejection forces help reduce the risk of Repetitive Strain Injury (RSI)
  • Super blow-out offers 150% increase in air boost to ensure efficient delivery of micro-volumes and prevent capillary action in 50 µl models and below
The pipettes work exclusively with Thermo Scientific ClipTip pipette tips, providing a unique tip interface that locks the tip in place, and ensures a complete seal with minimal tip application and ejection forces. Tips are locked firmly in place and will not fall off.
Thermo Scientific F1-ClipTip pipettes are warranted to be free from defects in material and workmanship for a product lifetime (10 years). This warranty is subject to conditions as stated in the attached warranty information document and require an online registration.
Leveringsinformation: Supplied with maintenance set (incl. grease and O-rings), tip fitting tool, ClipTip tip sample, shelf hanger, warning labels, calibration certificate, warranty certificate and user's guide (English, French, German, Spanish).