Finnpipette™ F1 Good Laboratory Pipetting (GLP) kits
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
4700850N 4700860N 4700865N 4701060N
613-5578EA 0 DKK
613-5578 613-5579 613-5580 613-5581
Finnpipette™ F1 Good Laboratory Pipetting (GLP) kits
Pipetter Automatiske pipetter
Finnpipette™ F1 GLP kits offer a variety of single channel pipettes in a range of volumes that are packaged with compatible Finntips, and a convenient pipette stand to support good laboratory pipetting in your lab.
  • Adjustable finger rest for increased ergonomics and comfortability
  • Set and forget pipetting button for easy and secured volume adjustment
  • Antimicrobial surface for better protection
  • Large easy to read display for better vision ergonomics
  • Soft-touch tip ejection for light end effortless tip ejection
Select the kit that is right for you, and take advantage of the Finnpipette™ F1 performance and lightweight design to maximize your comfort.
Warranty: 5 years with web registration.
Bestillingsoplysninger: The kit contains samples racks of Finntip Flex, the most advanced traditional tip offering very low attachment and ejection forces. The good laboratory pipetting guide contains helpful tips for pipetting including pipetting techniques, ergonomic considerations and how to select tips for best performance.
Leveringsinformation: All kits are delivered with F-stand, good laboratory pipetting guide, reagent reservoir demo pack.