Ship propeller roughness comparator scales
Leverandør: Industrial Physics
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
LD2041 LD2042
SIMELD2041EA 3400 DKK
SIMELD2041 SIMELD2042
Ship propeller roughness comparator scales
Belægnings tester
These roughness scales are designed to estimate the surface roughness, at touch or sight, specifically developed for the profiles related to the condition of ships propellers over life.
Bestillingsoplysninger: Each roughness scale includes six specimens.
Leveringsinformation: Supplied with guidance manual and recommended limits instructions.