Diluent, green
Leverandør: IMMUNOLOGIC
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
S/UD09-20 UD09-125 UD09-500 UD09-999
VWRKS/UD09-20EA 0 DKK
VWRKS/UD09-20 UD09-125 UD09-500 UD09-999
Diluent, green
Elektroforese reagenser Western Blotting-reagenser
Bright Diluent is provided as a ready to use normal antibody diluent intended for diluting and stabilising antibodies. It eliminates background encountered in immunohistochemistry staining procedures. The diluent contains complex proteins and non-protein mixtures which eliminates immunohistostaining background by blocking Fc receptors and prevents non-specific protein-protein interactions.
  • High-quality diluent solution for IHC applications
Certificeringer: Meets CE/IVD standards.
Leveringsinformation: The Immunologic products will be shipped with blue ice. Store the products between 2 to 8°C
Advarsel: Irritating to eyes, respiratory systems and skin.