Universal valve lockout, base clamping units
Leverandør: Brady
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
050924 050899
BRDY050924EA 816 DKK
BRDY050924 118-0515
Universal valve lockout, base clamping units
Sikkerhedslåse
These universal valve lockouts are effective for all types of standard valves.
  • Versatile, rugged, easy to use and lightweight
  • Can also be used to lock out large levers, T-handles and hard-to-secure mechanical devices
  • Open-ended clamp fits over closed rings and wide handles
  • Made of industrial-grade steel and nylon for extra impact and chemical resistance
Max. handle thickness: 15,24 mm
Max. valve handle width: 25,40 mm
Max. shackle diameter: 9,00 mm
Min. shackle clearance: 20,00 mm
Bestillingsoplysninger: Base unit only (cable and blocking arm sold separately).