Phosphate buffered saline solutions (PBS) without calcium and magnesium, HyClone™
Leverandør: HyClone products (Cytiva)
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SH30256.01 SH30258.01 SH30256.02 SH30256.FS SH30256.LS SH30258.02
SH30256.01EA 200 DKK
SH30256.01 HYCLSH30258.01 HYCLSH30256.02 HYCLSH30256.FS HYCLSH30256.LS HYCLSH30258.02
Phosphate buffered saline solutions (PBS) without calcium and magnesium, HyClone™
Buffere Vævskultur buffere
Balanced salt solution used for multiple cell culture applications.
  • Full traceability and documented origin of all formula ingredients
  • Validated and consistent production processes
0,1 µm sterile filtered
Endotoxin Concentration: ≤1.0 EU/mL
Certificeringer: Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities