Work positioning lanyard, Miller® HandZup®
Leverandør: HONEYWELL SAFETY
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1032097 1032098 1032099
113-8665EA 2040 DKK
113-8665 113-8666 113-8667
Work positioning lanyard, Miller® HandZup®
Faldbeskyttelse Faldliner og Tov
The ergonomic HandZup® is a unique, patented work positioning system (EN 358) that leaves the hands free and enables the operator to work in complete safety.
  • Easy to use, ensuring adjustment is quick, accurate and flexible for greater comfort while working
  • Miller HandZup® work positioning lanyard available in 2 m, 3 m or 4 m length
  • With a twist lock karabiner and SNAP HOOK and protective sheath
Certificeringer: EN 358