Fall protection harnesses, Miller H-Design® 2 pts harness Mating 2 loops
Leverandør: HONEYWELL SAFETY
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1032839 1032840 1032841
113-8651EA 1230 DKK
113-8651 113-8652 113-8653
Fall protection harnesses, Miller H-Design® 2 pts harness Mating 2 loops
Faldbeskyttelse Faldseler
Miller H-Design® two points harness with mating buckles on chest and legs straps and two sternal webbing loops. The Miller H-Design® harnesses are the new generation of harnesses dedicated to construction and general industry sectors providing more comfort, freedom of movements and safety to workers.
  • Straps are made from Duraflex Stretch and black webbing (PES with PTFE treatment)
  • Straps are water repellent and anti-stain treated
  • Sternal anchorage point with long webbing loops
  • Steel/zinc plated silver finished back anchorage D-ring and mating buckles
Weight: 1 kg
Breaking strength: >15 KN
Temperature: –30 to +50 °C
Certificeringer: Certified for a maximum load of 140 kg
ISO 9001/2000; EN 361; Cat III