Capillary columns, MXT®-65TG
Leverandør: Restek
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
77005 77007 77008 77010
REST77005EA 4670 DKK
REST77005 REST77007 REST77008 REST77010
Capillary columns, MXT®-65TG
Kromatografikolonner
MXT®-65TG columns are application-specific columns, specially tested for triglycerides.
  • High-polarity phase; Crossbond® diphenyl dimethyl polysiloxane
  • Stable to 370 °C
  • Coil diameter: 114 mm
The MXT®-65TG phase resolves triglycerides by degree of unsaturation as well as by carbon number. Because of the chemistry required to achieve 370 °C thermal stability, an MXT®-65TG column should not be used for analyses of compounds that contain active oxygenated groups.