Test sieves, synthetic
Leverandør: LINKER INDUSTRIE TECHN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
552050881-0025 552050881-0050 552050881-0080 552050881-1000 552050881-0100 552050881-0250 552050881-0036 552050881-0150 552050881-0063 552050881-0125 552050881-0200 552050881-0630
LINK552050881-0025EA 1130 DKK
LINK552050881-0025 LINK552050881-0050 LINK552050881-0080 LINK552050881-1000 LINK552050881-0100 LINK552050881-0250 LINK552050881-0036 LINK552050881-0150 LINK552050881-0063 LINK552050881-0125 LINK552050881-0200 LINK552050881-0630
Test sieves, synthetic
Sigter Testsigter
The synthetic POM frame, nylon mesh sieves meet DIN 4197 requirements and all current amendments.
  • Metal free construction