Low protein binding microcentrifuge tubes, Pierce™
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
90410 90411
PIER90410EA 274 DKK
PIER90410 PIER90411
Low protein binding microcentrifuge tubes, Pierce™
Rør Mikrocentrifugerør
PP, translucent.
  • Able to tolerate centrifuge speeds up to 20000 RCF
  • Suitable for IP and mass spectrometry applications
  • Certified RNase/DNase-free
Pierce™ low protein binding microcentrifuge tubes (1,5 ml) help minimise protein sample loss during sample manipulation and are ideal in applications where processing steps are extensive.