Sampling pouches, 400 series Kapak®
Leverandør: ProAmpac
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
400-24 406-24 408-24 400B-2M 400-F3-1MB 402-24 402B-2M 403-24 404-24 404B-2M 405B2-M 405-24
300-0294EA 319 DKK
300-0294 300-0304 300-0305 300-0296 300-0295 300-0297 300-0298 300-0299 300-0300 300-0301 300-0303 AMPA405-24
Sampling pouches, 400 series Kapak®
Poser Prøveposer
63 micron high barrier, crystal clear, three side seal, heat sealable polyester or foil pouches.
  • Pouches are microwaveable, boilable and freezable
  • High barrier leak proof pouches help eliminate cross contamination of dangerous substances
  • With 9 mm seals and tear notches
  • Infrared and gamma ray resistant
Typical applications: Medical, industrial, food preservation, veterinarian, health and beauty, agriculture.