General purpose gloves, DermiFlex Ultra Glove, A352
Leverandør: Portwest
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
A352G8RL A352G8RM A352G8RXL
112-4591EA 472 DKK
112-4591 112-4592 112-4593
General purpose gloves, DermiFlex Ultra Glove, A352
Handsker
These nylon/spandex gloves with PU/nitrile foam coating on palm and fingers are ideal for precision handling.
  • 3/4 coated for added protection to the back of the hand
  • Seamless breathable knitted liner for comfort and dexterity
EN 388 (3131)
Certificeringer: EN 388:2016 +A1:2018 - 4.1.3.1.X.