ESD safety shoes, lace-up, Sensation Up Low
Leverandør: ELTEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
72201/45 72201/46 72201/43 72201/44 72201/40 72201/41 72201/42 72201/39 72201/47
114-7347EA 619 DKK
114-7347 114-7348 114-7345 114-7346 114-7342 114-7343 114-7344 114-7341 114-7349
ESD safety shoes, lace-up, Sensation Up Low
Sko og støvler
Casual, vintage style ESD safety shoes for ladies made from hydrophobised textile material in 'denim look' and hydrophobised nubuck leather, a padded tongue plus a low profile, dual-density TPU/PU sole (Up).
  • Bioactive fabric lining
  • Replaceable insole; orthopaedic insoles possible
  • Full length inlay sole (ESD Pro), black
  • Metal-free, puncture resistant midsole
  • Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black/grey