Capillary columns, CP-Sil 88
Leverandør: VARIAN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
CP6173 CP6174 CP6172 CP6175
VARICP6173EA 17070 DKK
VARICP6173 VARICP6174 VARICP6172 VARICP6175
Capillary columns, CP-Sil 88
Kromatografikolonner
CP-Sil 88 columns have high selectivity towards positional and geometric isomers for ease-of-use.
  • Highly substituted cyanopropyl phase
  • Highest polarity, non-chemically bonded and stabilised