Calibration standards for thickness gauges
Leverandør: Industrial Physics
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
LD5418 LD5417 LD5400
SIMELD5418EA 679 DKK
SIMELD5418 SIMELD5417 SIMELD5400
Calibration standards for thickness gauges
Belægnings tester
Calibration standards are meant for verifying coating gauges - These plastic foils and shims are acting as standards.
  • Labelling with both metric and imperial units
Plastic shims are used for quick measurements when placed over metal to protect the probe from rough or hot surfaces.
A thickness gauge can measure shim coating over a smooth metal surface (zero plate). Certified plastic shims can be placed over an uncoated metal - Not suitable for use with magnetic pull-off gages.
Certificeringer: NIST
Emballering: In a plastic pouch.