Column Protection System
Leverandør: MACHEREY-NAGEL
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
718966 718967 718968 718969 718970 718971
554-5851EA 2230 DKK
554-5851 MANA718967 MANA718968 MANA718969 MANA718970 MANA718971
Column Protection System
Kromatografi forsyninger Kromatografi reservedele
The Column Protection System guard column holder is used for the installation of NUCLEODUR®, NUCLEOSIL®, NUCLEOSHELL® or UNIVERSAL RP guard columns (EC system) at analytical HPLC columns with ¹/₁₆" fittings.
  • Innovative and universal screw-on guard column holder system
  • Ideal protection for your analytical main column
  • Also suitable for ultra-fast HPLC due to minimised void volume
  • Special ferrules - pressure stability up to 1300 bar
It is advisable to replace the guard column if the silver membrane is contaminated (bright or dark discoloration). If theMore Product Information