Fuego SCS and Fuego SCS pro gas safety burners
Leverandør: WARTEWIG
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
8.200.000 8.204.000
WART8.200.000EA 5020 DKK
WART8.200.000 WART8.204.000
Fuego SCS and Fuego SCS pro gas safety burners
Varmeapparater Gasbrændere
Fuego SCS and Fuego SCS pro gas safety burners have the latest safety technology. Thanks to their innovative design and an extensive range of standard equipment, these laboratory gas burners are suitable for all applications that require a flame in the laboratory. The low profile housing allows you to work in comfort, and the streamlined design reduces the impact of air currents on clean room workbenches. The SCS (safety control system) uses the latest safety technology, constantly analysing potential dangers and triggering protective measures when necessary - such as the interruption of the gas supply. After prolonged use a residual heat indicator signals that the burner head is still hot, to help to prevent burns. Made entirely of stainless steel, and thanks to its fireproof controls and a display protected by special glass, the Fuego SCS can withstand even extreme laboratory conditions.
  • SCS (safety control system) and BHC (burner head control) which detects clogging
  • Conventional and exclusive DoubleClick IR sensor with adjustable IR sensor range
  • Six standard programmes for IR sensor and foot control with burning time display
  • Nozzles for natural gas and propane/butane
  • Can be operated wirelessly using the latest rechargeable battery technology (Fuego SCS pro model)
The burner can be connected to natural gas or propane/butane, and is suitable for use with a central gas supply or gas cartridges. The units will prompt user to obtain a replacement cartridge in advance of the supply running out. The easy-to-read display ensures you can always keep track of your work, and the language-less symbol menu navigation makes selecting functions quick and easy. The burner can either be operated with the foot control or the contact-free IR sensor. When switched on, the additional DoubleClick safety function allows you to activate the burner simply by switching on the IR sensor twice. The Fuego SCS pro model is also fitted with a tilt sensor and is enabled for wireless battery operation (two Li-ion batteries with charging station are optional accessories).
Certificeringer: Tested in accordance with DIN-DVGW; certificate NG-2211AS0167