Accessories and supplements for rapid translation system
Leverandør: biotechrabbit
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BR1401601 BR1401701 BR1401801
733-2336EA 3770 DKK
733-2336 733-2337 BTRABR1401801
Accessories and supplements for rapid translation system
Proteinekspressions Protein Ekspressionsreagenser
Accessories and supplements for Rapid Translation System (RTS).
RTS DnaK Supplement: E. coli chaperone to enhance correct folding and increase solubility even of large proteins
Enhance solubility of proteins expressed with RTS E. coli kits
Improve functional activity of proteins expressed with RTS E. coli kits
No substrate limit regarding protein size
Easily combine with RTS 100 and 500 E. coli platforms
RTS GroE Supplement: E. coli chaperone mix to enhance correct folding and increase solubility of expressed proteins
Enhance solubility of proteins expressed with RTS E. coli kits
Improve functional activity of proteins expressed with RTS E. coli kits
RTS Amino Acid Sampler: Set of 20 individual amino acids for amino acid specific protein labellng for structure determination
Ideal for labelling of proteins for NMR and X-Ray analyses
For use with RTS 500 and RTS 9000 E. coli kits