Rapid translation system (RTS), E. coli disulfide and fab kits
Leverandør: biotechrabbit
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BR1400401 BR1400501
733-2340EA 8880 DKK
733-2340 733-2343
Rapid translation system (RTS), E. coli disulfide and fab kits
Proteinekspressions Proteinekspressionssystemer
The Rapid Translation System (RTS) is a scalable in vitro protein expression platform that produces large amounts of protein for characterisation studies, functional assays, or structural analysis. RTS enables both transcription and translation utilising High-Yield (HY) and continuous-exchange, cell-free (CECF) technologies, with systems based on bacterial (E. coli) or eukaryotic (wheat germ) cell lysates. RTS offers the advantages of a cell-free protein expression system, whilst eliminating the need for laborious up and downstream steps typically associated with cell-based expression systems.
  • RTS 100 E. coli Disulfide Kit/RTS 500 E. coli Disulfide Kit: Microgram-scale protein expression of functional disulfide-bonded proteins
  • Express disulfide-bonded proteins in functional form due to the optimised lysate and reaction conditions
  • No refolding of aggregated proteins required
  • Express up to 80 μg protein per reaction, due to CECF technology