Rapid translation system (RTS), insect membrane kits
Leverandør: biotechrabbit
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BR1401501 BR1401502
BTRABR1401501EA 2530 DKK
BTRABR1401501 BTRABR1401502
Rapid translation system (RTS), insect membrane kits
Proteinekspressions Proteinekspressionssystemer
RTS 100 Insect Membrane Kit provides high yields of in vitro synthesised post-translationally modified eukaryotic proteins.
  • Robust expression of a wide range of proteins with a range of modifications (clotting factors, protein kinases, and transcription factors)
  • Expression of membrane proteins without the need for additives
  • 3-fold higher yields of up to 50 µg/ml for enough protein for functional studies/screening procedures
  • Fast screening and increased flexibility using PCR products or circular plasmids as templates
The Rapid Translation System (RTS) is a scalable in vitro protein expression platform that produces large amounts of protein for characterisation studies, functional assays, or structural analysis. RTS enables both transcription and translation utilising High-Yield (HY) and continuous-exchange, cell-free (CECF) technologies, with systems based on bacterial (E. coli) or eukaryotic (wheat germ) cell lysates. RTS offers the advantages of a cell-free protein expression system, whilst eliminating the need for laborious up and downstream steps typically associated with cell-based expression systems.