Test kits, photometric, copper, LCK cuvette tests
Leverandør: Hach
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
LCK329 LCK529 LCK229
705-3026EA 1370 DKK
705-3026 705-3027 HACHLCK229
Test kits, photometric, copper, LCK cuvette tests
Miljø test kit
Cuvette tests for water testing applications.
  • Precise and reliable measurement values
  • Maximum safety for users, thanks to the closed system and low amounts of reagents
  • Convenient and error-free dosing of the reagents
  • Barcode label for automatic recognition in the photometer
  • Differentiation between tests and measuring ranges by means of colour-coding
Storage conditions: 2 to 8 °C (keep refrigerated) for LCK529 and 15 to 25 °C for LCK229 and LCK329
Information om tilbehør: Cuvette tests are officially approved for many legally required consent limits, with the help of Addista standard solutions and round-robin test solutions, they provide the assurance you need.